Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293640.1 | NP_001280569.1 | 24, 25 | 2593 | ||
NM_015275.2 | NP_056090.1 | 24, 25 | 2590 | |||
XM_011538074.2 | XP_011536376.1 | 24, 25 | 2556 | |||
XM_017019082.1 | XP_016874571.1 | 24, 25 | 2590 | |||
GenBank mRNA | AB028956.1 | - | 24, 25 | 2572 | ||
AK001728.1 | - | 24, 25 | 2557 | |||
AK023146.1 | - | 21, 22 | 2225 | |||
AK026950.1 | - | 13, 14 | 1512 | |||
AL137753.1 | - | 21, 22 | 2214 | |||
BC040936.1 | - | 1, 2 | 104 | |||
BC104992.1 | - | 24, 25 | 2532 | 8143995 | ||
BC104994.1 | - | 24, 25 | 2532 | 8143997 | ||
BC110850.1 | - | 24, 25 | 2541 | |||
BC143373.1 | - | 24, 25 | 2535 | 9051878 |