Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185075.1 | NP_001172004.1 | 15, 16 | 1703 | ||
NM_001185076.1 | NP_001172005.1 | 15, 16 | 1911 | |||
NM_001185081.1 | NP_001172010.1 | 14, 15 | 1640 | |||
NM_001185082.1 | NP_001172011.1 | 15, 16 | 1836 | |||
NM_002024.5 | NP_002015.1 | 16, 17 | 1974 | |||
NR_033699.1 | 15, 16 | 1778 | ||||
NR_033700.1 | 14, 15 | 1715 | ||||
GenBank mRNA | AB209188.1 | - | 11, 12 | 2401 | ||
AK294104.1 | - | 5, 6 | 619 | |||
BC086957.1 | - | 15, 16 | 1866 | 30347992 | ||
CX871385.1 | - | 4, 5 | 549 | |||
KC774028.1 | - | 14, 15 | 1382 | |||
KF856234.1 | - | 14, 15 | 1573 | |||
KJ534836.1 | - | 15, 16 | 1682 | |||
KJ534837.1 | - | 16, 17 | 1745 | |||
KM091457.1 | - | 17, 18 | 1853 | |||
KM091458.1 | - | 16, 17 | 1790 | |||
S65791.1 | - | 16, 17 | 1964 | |||
X69962.1 | - | 16, 17 | 1964 |