Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145418.1 | NP_001138890.1 | 8 | 2994 | ||
XM_005261405.2 | XP_005261462.1 | 8 | 2994 | |||
XM_006724171.3 | XP_006724234.1 | 7 | 11772 | |||
XM_011530018.2 | XP_011528320.1 | 8 | 3173 | |||
XM_011530019.2 | XP_011528321.1 | 8 | 2994 | |||
XM_011530020.1 | XP_011528322.1 | 8 | 2994 | |||
XM_011530021.2 | XP_011528323.1 | 8 | 2994 | |||
XM_011530022.1 | XP_011528324.1 | 8 | 2994 | |||
XM_017028673.1 | XP_016884162.1 | 8 | 2967 | |||
GenBank mRNA | AB028966.3 | - | 3 | 1787 | ||
AB665284.1 | - | 8 | 2942 |