Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPY19L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015283.1 | NP_056098.1 | 21 | 1945 | ||
XM_011515245.2 | XP_011513547.1 | 21 | 2094 | |||
XM_011515246.2 | XP_011513548.1 | 20 | 2007 | |||
GenBank mRNA | AB020684.1 | - | 18 | 1516 | ||
AJ011911.1 | - | 2 | 164 | |||
BC029591.1 | - | 2 | 206 | |||
DQ287932.1 | - | 21 | 1945 |