Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318339.1 | NP_001305268.1 | 3 | 521 | ||
NM_015308.3 | NP_056123.2 | 3 | 515 | |||
GenBank mRNA | AB023231.2 | - | 3 | 514 | ||
AK054636.1 | - | 3 | 338 | |||
AL137480.1 | - | 3 | 314 | |||
BC037404.1 | - | 3 | 346 | 5140963 | ||
DA238225.1 | - | 3 | 503 |