Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173431.1 | NP_001166902.1 | 10 | 3675 | ||
NM_015311.2 | NP_056126.1 | 10 | 3675 | |||
XM_005246424.4 | XP_005246481.1 | 10 | 3699 | |||
XM_011510857.2 | XP_011509159.1 | 10 | 3702 | |||
XM_011510863.2 | XP_011509165.1 | 10 | 3694 | |||
XM_011510864.2 | XP_011509166.1 | 10 | 3694 | |||
XM_011510865.2 | XP_011509167.1 | 10 | 3694 | |||
XM_017003696.1 | XP_016859185.1 | 10 | 3701 | |||
XM_017003697.1 | XP_016859186.1 | 10 | 3701 | |||
XM_017003698.1 | XP_016859187.1 | 10 | 3700 | |||
GenBank mRNA | AK023854.1 | - | 2 | 342 | ||
EF063637.1 | - | 10 | 3421 | |||
EF063638.1 | - | 10 | 3418 |