Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301084.1 | NP_001288013.1 | 4, 5 | 356 | ||
NM_015313.2 | NP_056128.1 | 4, 5 | 530 | |||
XM_006718805.3 | XP_006718868.1 | 4, 5 | 889 | |||
XM_011542720.2 | XP_011541022.1 | 4, 5 | 471 | |||
XM_017017421.1 | XP_016872910.1 | 4, 5 | 746 | |||
XM_017017422.1 | XP_016872911.1 | 4, 5 | 358 | |||
GenBank mRNA | AF180681.1 | - | 4, 5 | 202 | ||
AK294803.1 | - | 4, 5 | 331 | |||
BX647785.1 | - | 4, 5 | 332 | |||
BX648515.1 | - | 4, 5 | 337 | |||
DC350360.1 | - | 4, 5 | 356 |