Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098482.1 | NP_001091952.1 | 13 | 1600 | ||
NM_015321.2 | NP_056136.2 | 12 | 1552 | |||
XM_005259835.3 | XP_005259892.1 | 13 | 1302 | |||
XM_005259836.3 | XP_005259893.1 | 12 | 1197 | |||
XM_006722710.3 | XP_006722773.1 | 11 | 1146 | |||
XM_011527842.2 | XP_011526144.1 | 12 | 1456 | |||
XM_017026536.1 | XP_016882025.1 | 13 | 1561 | |||
GenBank mRNA | AB014516.1 | - | 13 | 1465 | ||
AY360171.1 | - | 13 | 1537 | |||
BC017075.2 | - | 8 | 1033 | |||
BC023614.2 | - | 12 | 1375 | |||
BC028050.1 | - | 12 | 1484 | 4938995 |