Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 22 | 3468 | ||
NM_001281749.1 | NP_001268678.1 | 21 | 3344 | |||
NM_025164.4 | NP_079440.3 | 21 | 3524 | |||
XM_005271481.2 | XP_005271538.1 | 22 | 3706 | |||
XM_005271482.4 | XP_005271539.2 | 21 | 3562 | |||
XM_005271484.3 | XP_005271541.2 | 21 | 3382 | |||
XM_005271485.3 | XP_005271542.1 | 9 | 2044 | |||
XM_011542721.2 | XP_011541023.1 | 22 | 3668 | |||
XM_011542722.1 | XP_011541024.1 | 22 | 3526 | |||
XM_011542723.2 | XP_011541025.1 | 22 | 3549 | |||
XM_011542724.2 | XP_011541026.1 | 21 | 3403 | |||
XM_011542725.2 | XP_011541027.1 | 20 | 3286 | |||
XM_017017424.1 | XP_016872913.1 | 20 | 3238 | |||
XM_017017425.1 | XP_016872914.1 | 22 | 3744 | |||
XM_017017426.1 | XP_016872915.1 | 20 | 3618 | |||
XM_017017427.1 | XP_016872916.1 | 17 | 3021 | |||
GenBank mRNA | AB209327.1 | - | 14 | 2534 | ||
AK022302.1 | - | 3 | 1079 | |||
AK122838.1 | - | 7 | 2786 | |||
AY598338.1 | - | 3 | 1076 | |||
BC063887.1 | - | 7 | 1537 | |||
BC086858.1 | - | 3 | 1079 | |||
BC128510.1 | - | 21 | 3206 | 40081639 | ||
BC128511.1 | - | 22 | 3324 | 40081640 |