Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 24 | 3896 | ||
NM_001281749.1 | NP_001268678.1 | 23 | 3772 | |||
NM_025164.4 | NP_079440.3 | 23 | 3952 | |||
XM_005271481.2 | XP_005271538.1 | 24 | 4134 | |||
XM_005271482.4 | XP_005271539.2 | 23 | 3990 | |||
XM_005271484.3 | XP_005271541.2 | 23 | 3810 | |||
XM_005271485.3 | XP_005271542.1 | 11 | 2472 | |||
XM_011542721.2 | XP_011541023.1 | 24 | 4096 | |||
XM_011542722.1 | XP_011541024.1 | 24 | 3954 | |||
XM_011542723.2 | XP_011541025.1 | 24 | 3977 | |||
XM_011542724.2 | XP_011541026.1 | 23 | 3831 | |||
XM_011542725.2 | XP_011541027.1 | 22 | 3714 | |||
XM_017017424.1 | XP_016872913.1 | 22 | 3666 | |||
XM_017017425.1 | XP_016872914.1 | 24 | 4172 | |||
XM_017017426.1 | XP_016872915.1 | 22 | 4046 | |||
XM_017017427.1 | XP_016872916.1 | 19 | 3449 | |||
GenBank mRNA | AB209327.1 | - | 16 | 2962 | ||
AI765994.1 | - | 2 | 352 | |||
AK022302.1 | - | 5 | 1507 | |||
AK122838.1 | - | 9 | 3214 | |||
AY598338.1 | - | 5 | 1504 | |||
BC063887.1 | - | 9 | 1965 | |||
BC086858.1 | - | 5 | 1507 | |||
BC128510.1 | - | 23 | 3634 | 40081639 | ||
BC128511.1 | - | 24 | 3752 | 40081640 | ||
BM682365.1 | - | 2 | 284 |