Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 7 | 872 | ||
NR_033698.1 | 6 | 786 | ||||
GenBank mRNA | AB209689.1 | - | 1 | 6536 | ||
AK091717.1 | - | 2 | 1715 | |||
AK123002.1 | - | 7 | 1259 | |||
AK292121.1 | - | 7 | 848 | |||
BC017029.2 | - | 7 | 815 | |||
BC084566.1 | - | 7 | 832 | 6423445 | ||
BT009854.1 | - | 7 | 824 | |||
BX648381.1 | - | 2 | 2643 | |||
L00634.1 | - | 7 | 840 | |||
L10413.1 | - | 7 | 830 |