Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXOSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282708.1 | NP_001269637.1 | 7 | 656 | ||
NM_001282709.1 | NP_001269638.1 | 7 | 644 | |||
NM_014285.6 | NP_055100.2 | 8 | 734 | |||
NR_104230.1 | 7 | 1240 | ||||
XM_005272176.2 | XP_005272233.1 | 8 | 732 | |||
XM_006717023.2 | XP_006717086.1 | 7 | 642 | |||
XM_017014558.1 | XP_016870047.1 | 7 | 1266 | |||
GenBank mRNA | AK001916.1 | - | 8 | 723 | ||
AK022460.1 | - | 8 | 720 | |||
AK090512.1 | - | 7 | 1266 | |||
AK092727.1 | - | 2 | 1500 | |||
AK296605.1 | - | 7 | 645 | |||
AK303163.1 | - | 7 | 1220 | |||
AL573310.3 | - | 2 | 531 | |||
BC000747.2 | - | 8 | 704 | 2821702 | ||
BC128445.1 | - | 2 | 88 |