Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142498.1 | NP_001135970.1 | 8 | 1588 | ||
NM_001314049.1 | NP_001300978.1 | 7 | 1377 | |||
NM_012238.4 | NP_036370.2 | 9 | 2094 | |||
GenBank mRNA | AF083106.2 | - | 9 | 2094 | ||
AF235040.1 | - | 9 | 2089 | |||
AK027686.1 | - | 6 | 1151 | |||
AK074805.1 | - | 7 | 1295 | |||
AK289743.1 | - | 9 | 1830 | |||
AL136741.1 | - | 9 | 1709 | |||
BC012499.1 | - | 9 | 1674 | 4518906 | ||
BX648554.1 | - | 6 | 1337 |