Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178074.1 | NP_001171545.1 | 12 | 1670 | ||
NM_021150.3 | NP_066973.2 | 12 | 1670 | |||
XM_005268754.4 | XP_005268811.1 | 13 | 1746 | |||
XM_005268757.4 | XP_005268814.1 | 12 | 1665 | |||
XM_011538093.2 | XP_011536395.1 | 13 | 1799 | |||
XM_017019098.1 | XP_016874587.1 | 13 | 2095 | |||
XM_017019099.1 | XP_016874588.1 | 13 | 4754 | |||
XM_017019100.1 | XP_016874589.1 | 12 | 1939 | |||
XM_017019101.1 | XP_016874590.1 | 12 | 4598 | |||
XM_017019102.1 | XP_016874591.1 | 12 | 4595 | |||
GenBank mRNA | AJ133439.1 | - | 6 | 793 | ||
AK095519.1 | - | 12 | 1670 | |||
AK295565.1 | - | 11 | 1476 | |||
AK307576.1 | - | 12 | 1508 | |||
BC029871.1 | - | 10 | 1234 | |||
BC115393.1 | - | 12 | 1498 | 40053971 | ||
BC115394.1 | - | 12 | 1498 | 40053972 |