Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014297.4 | NP_055112.2 | 3 | 324 | ||
XM_005258687.3 | XP_005258744.1 | 3 | 421 | |||
GenBank mRNA | AI879202.1 | - | 3 | 322 | ||
AK309880.1 | - | 3 | 269 | |||
AK315346.1 | - | 3 | 287 | |||
BC008250.1 | - | 3 | 274 | 3872059 | ||
BP252912.1 | - | 3 | 324 | |||
D83198.2 | - | 3 | 313 | |||
KU178565.1 | - | 3 | 245 |