Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243225.1 | NP_001230154.1 | 9 | 1002 | ||
NM_001282327.1 | NP_001269256.1 | 11 | 1400 | |||
NM_001282328.1 | NP_001269257.1 | 11 | 1447 | |||
NM_014303.3 | NP_055118.1 | 9 | 1002 | |||
XM_006724173.1 | XP_006724236.1 | 9 | 958 | |||
XM_017028678.1 | XP_016884167.1 | 6 | 2820 | |||
GenBank mRNA | AB451380.1 | - | 9 | 895 | ||
AK096320.1 | - | 11 | 1419 | |||
AK127185.1 | - | 11 | 1401 | |||
AK315518.1 | - | 9 | 969 | |||
BC032489.1 | - | 9 | 959 | 5416478 | ||
CR456539.1 | - | 9 | 920 | |||
U78310.1 | - | 9 | 953 |