Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 32 | 6076 | ||
XM_017021124.1 | XP_016876613.1 | 32 | 6095 | |||
XM_017021125.1 | XP_016876614.1 | 32 | 6095 | |||
XM_017021126.1 | XP_016876615.1 | 31 | 6709 | |||
XM_017021127.1 | XP_016876616.1 | 21 | 3791 | |||
XM_017021128.1 | XP_016876617.1 | 21 | 3644 | |||
GenBank mRNA | AB002319.1 | - | 17 | 2946 | ||
AB425197.1 | - | 32 | 6064 | |||
AK024743.1 | - | 4 | 438 | |||
BC144594.1 | - | 30 | 5380 | |||
BX537886.1 | - | 32 | 6070 | |||
BX538025.1 | - | 28 | 5569 | |||
BX648683.1 | - | 32 | 6074 | |||
CR749276.1 | - | 33 | 6011 | |||
CR749365.1 | - | 32 | 6075 |