Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015346.3 | NP_056161.2 | 37 | 7103 | ||
XM_017021124.1 | XP_016876613.1 | 37 | 7122 | |||
XM_017021125.1 | XP_016876614.1 | 37 | 7122 | |||
XM_017021126.1 | XP_016876615.1 | 36 | 7736 | |||
XM_017021127.1 | XP_016876616.1 | 26 | 4818 | |||
XM_017021128.1 | XP_016876617.1 | 26 | 4671 | |||
GenBank mRNA | AB002319.1 | - | 22 | 3973 | ||
AB425197.1 | - | 37 | 7091 | |||
AK024743.1 | - | 9 | 1465 | |||
BC008927.2 | - | 3 | 502 | |||
BC033235.1 | - | 4 | 798 | |||
BC144594.1 | - | 35 | 6407 | |||
BX537886.1 | - | 37 | 7097 | |||
BX538025.1 | - | 33 | 6596 | |||
CR749276.1 | - | 38 | 7038 | |||
CR749365.1 | - | 36 | 7716 |