Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134476.1 | NP_001127948.1 | 7 | 554 | ||
NM_015350.2 | NP_056165.1 | 5 | 611 | |||
XM_005270701.4 | XP_005270758.1 | 5 | 480 | |||
XM_005270702.4 | XP_005270759.1 | 6 | 501 | |||
XM_005270703.4 | XP_005270760.1 | 5 | 2248 | |||
XM_011541143.2 | XP_011539445.1 | 6 | 538 | |||
XM_011541144.2 | XP_011539446.1 | 7 | 850 | |||
XM_011541145.2 | XP_011539447.1 | 8 | 875 | |||
XM_011541146.2 | XP_011539448.1 | 6 | 2305 | |||
XM_011541148.2 | XP_011539450.1 | 6 | 526 | |||
XM_017000883.1 | XP_016856372.1 | 6 | 1500 | |||
XM_017000884.1 | XP_016856373.1 | 5 | 2402 | |||
XM_017000885.1 | XP_016856374.1 | 5 | 572 | |||
XM_017000886.1 | XP_016856375.1 | 5 | 476 | |||
GenBank mRNA | AF385436.1 | - | 5 | 611 | ||
AY358112.1 | - | 4 | 362 | |||
BC053565.1 | - | 1 | 2218 | |||
D86984.2 | - | 5 | 917 |