Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK8IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012324.4 | NP_036456.1 | 9 | 2308 | ||
XM_011530679.2 | XP_011528981.1 | 9 | 2200 | |||
XM_011530680.2 | XP_011528982.1 | 9 | 2086 | |||
XM_011530681.2 | XP_011528983.1 | 9 | 2065 | |||
GenBank mRNA | AF136382.1 | - | 9 | 2291 | ||
AF218778.1 | - | 7 | 2093 | |||
AK293580.1 | - | 7 | 679 | |||
AK294265.1 | - | 10 | 1367 | |||
AL021708.1 | - | 5 | 1311 | |||
BC009940.2 | - | 8 | 1058 | 4100637 | ||
BC047527.1 | - | 9 | 2221 | 5277237 | ||
U79261.1 | - | 4 | 382 |