Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012341.2 | NP_036473.2 | 7, 8 | 928 | ||
GenBank mRNA | AF120334.1 | - | 7, 8 | 880 | ||
AF325353.1 | - | 7, 8 | 857 | |||
AK001548.1 | - | 7, 8 | 875 | |||
AK001552.1 | - | 7, 8 | 930 | |||
AK097093.1 | - | 5, 6 | 1736 | |||
AK222861.1 | - | 7, 8 | 875 | |||
AK295231.1 | - | 6, 7 | 771 | |||
AK301721.1 | - | 7, 8 | 1043 | |||
AK302219.1 | - | 6, 7 | 733 | |||
BC033784.1 | - | 7, 8 | 851 | |||
BC038975.2 | - | 7, 8 | 889 | 5534986 |