Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014314.3 | NP_055129.2 | 8 | 1210 | ||
GenBank mRNA | AF038963.1 | - | 8 | 1209 | ||
AK125989.1 | - | 9 | 1415 | |||
AK301678.1 | - | 7 | 933 | |||
AK315040.1 | - | 8 | 1081 | |||
BC132786.1 | - | 8 | 1158 | 40146808 | ||
BC136610.1 | - | 8 | 1158 | 9020600 | ||
BX647917.1 | - | 7 | 947 |