Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM89B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098784.1 | NP_001092254.1 | 2 | 876 | ||
NM_001098785.1 | NP_001092255.1 | 2 | 866 | |||
NM_152832.2 | NP_690045.1 | 2 | 827 | |||
GenBank mRNA | AF052151.1 | - | 2 | 763 | ||
BC023991.2 | - | 3 | 1274 | 2819240 | ||
BC032373.1 | - | 2 | 838 | 5123262 | ||
BE394215.1 | - | 2 | 409 | |||
BQ927525.1 | - | 1 | 67 | |||
DR001553.1 | - | 2 | 663 |
Gene Symbol: MTVR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027025.1 | 1 | 249 |