Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130106.1 | NP_001123578.1 | 5, 6 | 973 | ||
NM_001297600.1 | NP_001284529.1 | 3, 4 | 888 | |||
NM_012267.4 | NP_036399.3 | 4, 5 | 1082 | |||
XM_005258700.2 | XP_005258757.1 | 4, 5 | 813 | |||
XM_005258701.2 | XP_005258758.1 | 4, 5 | 875 | |||
XM_011526691.1 | XP_011524993.1 | 5, 6 | 971 | |||
XM_017026543.1 | XP_016882032.1 | 4, 5 | 810 | |||
XM_017026544.1 | XP_016882033.1 | 4, 5 | 810 | |||
XM_017026545.1 | XP_016882034.1 | 4, 5 | 813 | |||
GenBank mRNA | AB020592.1 | - | 4, 5 | 804 | ||
AF093420.1 | - | 4, 5 | 807 | |||
AF187859.1 | - | 4, 5 | 772 | |||
AF217996.1 | - | 5, 6 | 959 | |||
AK075293.1 | - | 4, 5 | 829 | |||
AK129755.1 | - | 4, 5 | 885 | |||
AK130781.1 | - | 4, 5 | 819 | |||
AK294358.1 | - | 3, 4 | 887 | |||
BC001236.1 | - | 3, 4 | 737 | 3454675 | ||
BC002373.2 | - | 4, 5 | 794 | 2822836 | ||
CR457118.1 | - | 3, 4 | 639 |