Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018070.4 | NP_060540.2 | 6 | 806 | ||
NM_145716.3 | NP_663768.1 | 6 | 806 | |||
XM_017000897.1 | XP_016856386.1 | 6 | 475 | |||
XM_017000898.1 | XP_016856387.1 | 3 | 702 | |||
GenBank mRNA | AF500116.1 | - | 6 | 634 | ||
AK001217.1 | - | 2 | 154 | |||
AL519196.3 | - | 6 | 781 | |||
BC003605.1 | - | 6 | 445 | 3546126 | ||
BC066365.1 | - | 6 | 550 | 6169677 |