Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032012.3 | NP_114401.2 | 17 | 2507 | ||
XM_011518446.2 | XP_011516748.1 | 17 | 2522 | |||
XM_011518447.1 | XP_011516749.1 | 16 | 2483 | |||
XM_011518449.1 | XP_011516751.1 | 16 | 2390 | |||
XM_011518451.1 | XP_011516753.1 | 15 | 2378 | |||
XM_011518452.1 | XP_011516754.1 | 16 | 2273 | |||
XM_017014571.1 | XP_016870060.1 | 16 | 2480 | |||
XM_017014572.1 | XP_016870061.1 | 15 | 2249 | |||
GenBank mRNA | AF153415.1 | - | 17 | 2507 | ||
AK074248.1 | - | 7 | 805 | |||
AK090543.1 | - | 3 | 1562 | |||
AK172815.1 | - | 11 | 1252 | |||
AK299952.1 | - | 17 | 2489 | |||
AL833960.1 | - | 15 | 1898 | |||
BC014248.2 | - | 7 | 858 |