Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033054.2 | NP_001028226.1 | 3 | 460 | ||
NM_001033055.2 | NP_001028227.1 | 3 | 469 | |||
NM_001285399.2 | NP_001272328.1 | 4 | 613 | |||
NM_001285400.2 | NP_001272329.1 | 4 | 583 | |||
NM_001285401.2 | NP_001272330.1 | 4 | 649 | |||
NM_001285402.1 | NP_001272331.1 | 4 | 640 | |||
NM_001285403.2 | NP_001272332.1 | 4 | 625 | |||
NM_014336.4 | NP_055151.3 | 4 | 649 | |||
GenBank mRNA | AB593042.1 | - | 4 | 546 | ||
AB593043.1 | - | 4 | 570 | |||
AB593044.1 | - | 4 | 584 | |||
AB593045.1 | - | 4 | 584 | |||
AB593046.1 | - | 4 | 586 | |||
AB593047.1 | - | 4 | 627 | |||
AB593048.1 | - | 4 | 640 | |||
AB593049.1 | - | 4 | 594 | |||
AB593050.1 | - | 4 | 570 | |||
AB593051.1 | - | 4 | 571 | |||
AB593052.1 | - | 4 | 570 | |||
AB593053.1 | - | 3 | 426 | |||
AB593054.1 | - | 3 | 369 | |||
AB593055.1 | - | 4 | 615 | |||
AF038437.1 | - | 4 | 568 | |||
AF148864.1 | - | 4 | 734 | |||
AF525970.1 | - | 3 | 460 | |||
AJ633677.1 | - | 3 | 422 | |||
AJ633678.1 | - | 3 | 413 | |||
AJ830742.1 | - | 4 | 537 | |||
AJ830743.1 | - | 4 | 603 | |||
AK023970.1 | - | 4 | 577 | |||
AK314558.1 | - | 4 | 570 | |||
BC012055.1 | - | 4 | 562 | 4634156 | ||
BX537907.1 | - | 4 | 547 | |||
BX647279.1 | - | 4 | 615 |