Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PISD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_001326413.1 | NP_001313342.1 | 3 | 700 | ||
NM_001326414.1 | NP_001313343.1 | 4 | 780 | |||
NM_001326415.1 | NP_001313344.1 | 4 | 954 | |||
NM_001326416.1 | NP_001313345.1 | 6 | 1143 | |||
NM_001326417.1 | NP_001313346.1 | 5 | 1034 | |||
NM_001326419.1 | NP_001313348.1 | 5 | 1034 | |||
NM_014338.3 | NP_055153.1 | 4 | 586 | |||
NM_178022.1 | NP_821141.1 | 3 | 495 | |||
GenBank mRNA | AY927618.1 | - | LIT3606 AY927618 |
2 | 155 | |
BC001482.2 | - | 4 | 586 | 3508281 |