Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 1790 | ||
NM_198391.2 | NP_938205.1 | 3 | 1984 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 2502 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 2639 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 2301 | |||
GenBank mRNA | AB040902.1 | - | 2 | 1678 | ||
AF169677.1 | - | 2 | 1687 | |||
AK027297.1 | - | 2 | 1701 | |||
AK027670.1 | - | 1 | 898 | |||
AK027694.1 | - | 1 | 1419 | |||
AK074883.1 | - | 2 | 1731 | |||
AK074909.1 | - | 2 | 1659 | |||
AY358319.1 | - | 1 | 1524 | |||
BC020870.1 | - | 2 | 1697 | 4606852 |