Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRZB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001463.3 | NP_001454.2 | 3 | 787 | ||
GenBank mRNA | AK130009.1 | - | 4 | 439 | ||
AK312741.1 | - | 3 | 666 | |||
BC027855.1 | - | 3 | 639 | 5221358 | ||
BT019883.1 | - | 3 | 569 | |||
U24163.1 | - | 3 | 777 | |||
U68057.1 | - | 3 | 629 | |||
U91903.1 | - | 3 | 638 |
Set Membership: |
G-protein coupled receptor |