Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170634.1 | NP_001164105.1 | 6 | 644 | ||
NM_001170937.1 | NP_001164408.1 | 6 | 635 | |||
NM_004960.3 | NP_004951.1 | 6 | 647 | |||
NR_028388.2 | 6 | 647 | ||||
XM_005255233.4 | XP_005255290.1 | 6 | 1310 | |||
XM_011545781.1 | XP_011544083.1 | 6 | 613 | |||
XM_011545782.2 | XP_011544084.1 | 2 | 38 | |||
GenBank mRNA | AB208902.1 | - | 6 | 607 | ||
AK098774.1 | - | 6 | 615 | |||
AK130774.1 | - | 6 | 619 | |||
AK290936.1 | - | 6 | 618 | |||
BC000402.2 | - | 6 | 610 | 2822692 | ||
BC002459.1 | - | 6 | 582 | 3345294 | ||
BC026062.2 | - | 6 | 600 | 3937437 | ||
BT007131.1 | - | 5 | 542 | |||
BX423519.2 | - | 6 | 626 | |||
CR456747.1 | - | 5 | 542 | |||
GU933437.1 | - | 2 | 220 | |||
S62140.1 | - | 6 | 620 | |||
X71428.1 | - | 6 | 611 |