Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122951.2 | NP_001116423.1 | 1 | 257 | ||
NM_002036.3 | NP_002027.2 | 2 | 1023 | |||
GenBank mRNA | AF030521.1 | - | 1 | 82 | ||
AK291593.1 | - | 2 | 1023 | |||
AK309521.1 | - | 1 | 526 | |||
AY167991.1 | - | 1 | 76 | |||
BC017817.1 | - | 2 | 228 | 4696425 | ||
BC095496.1 | - | 1 | 79 | |||
BX537430.1 | - | 3 | 920 | |||
CD701182.1 | - | 2 | 337 | |||
EL735542.1 | - | 2 | 76 | |||
HE572751.1 | - | 2 | 76 | |||
JX081310.1 | - | 1 | 82 | |||
U01839.1 | - | 1 | 257 |