Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243093.1 | NP_001230022.1 | 2 | 1156 | ||
NM_001465.4 | NP_001456.3 | 2 | 1203 | |||
NM_199335.3 | NP_955367.1 | 2 | 1203 | |||
XM_006714464.2 | XP_006714527.1 | 2 | 1188 | |||
XM_006714465.2 | XP_006714528.1 | 3 | 1541 | |||
XM_006714466.2 | XP_006714529.1 | 2 | 1317 | |||
XM_011514008.2 | XP_011512310.1 | 4 | 1641 | |||
XM_011514009.1 | XP_011512311.1 | 4 | 1562 | |||
XM_011514010.1 | XP_011512312.1 | 3 | 1280 | |||
XM_011514011.2 | XP_011512313.1 | 2 | 1164 | |||
XM_011514012.2 | XP_011512314.1 | 2 | 1164 | |||
XM_011514013.2 | XP_011512315.1 | 2 | 1165 | |||
XM_017009316.1 | XP_016864805.1 | 2 | 1163 | |||
GenBank mRNA | AF001862.1 | - | 1 | 1107 | ||
AF198052.1 | - | 1 | 1040 | |||
AK130159.1 | - | 2 | 1198 | |||
AK290403.1 | - | 2 | 1198 | |||
AK297077.1 | - | 2 | 1156 | |||
BC015933.1 | - | 2 | 1132 | |||
BC117449.1 | - | 2 | 1102 | 40126000 | ||
BC143645.1 | - | 2 | 1102 | 9052159 | ||
BX647195.1 | - | 2 | 1188 | |||
BX647196.1 | - | 2 | 1189 | |||
U93049.1 | - | 1 | 1070 |