Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002037.5 | NP_002028.1 | 4 | 687 | ||
NM_153047.3 | NP_694592.1 | 1 | 91 | |||
NM_153048.3 | NP_694593.1 | 2 | 183 | |||
XM_005266890.3 | XP_005266947.1 | 4 | 268 | |||
XM_005266892.3 | XP_005266949.1 | 4 | 268 | |||
XM_017010650.1 | XP_016866139.1 | 3 | 425 | |||
XM_017010651.1 | XP_016866140.1 | 3 | 329 | |||
XM_017010652.1 | XP_016866141.1 | 2 | 339 | |||
XM_017010653.1 | XP_016866142.1 | 2 | 183 | |||
XM_017010654.1 | XP_016866143.1 | 2 | 164 | |||
XM_017010655.1 | XP_016866144.1 | 4 | 268 | |||
GenBank mRNA | AB451293.1 | - | 1 | 80 | ||
AB451426.1 | - | 1 | 80 | |||
AK056699.1 | - | 3 | 420 | |||
BC032496.1 | - | 2 | 134 | 5219761 | ||
CX781584.1 | - | 3 | 344 | |||
CX872011.1 | - | 2 | 121 | |||
DA815564.1 | - | 2 | 184 | |||
DA922886.1 | - | 2 | 178 | |||
M14333.1 | - | 4 | 659 | |||
M14676.1 | - | 4 | 450 | |||
S74774.1 | - | 1 | 80 |