Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010875.3 | NP_001010875.1 | 10 | 1073 | ||
NM_001286806.1 | NP_001273735.1 | 9 | 954 | |||
NM_001286807.1 | NP_001273736.1 | 10 | 1131 | |||
XM_005266321.2 | XP_005266378.1 | 9 | 1221 | |||
XM_011535027.2 | XP_011533329.1 | 9 | 850 | |||
XM_017020524.1 | XP_016876013.1 | 10 | 1123 | |||
GenBank mRNA | AK094133.1 | - | 10 | 1000 | ||
AK095465.1 | - | 9 | 853 | |||
AK302967.1 | - | 9 | 881 | |||
AL832206.1 | - | 4 | 3371 | |||
BC132739.1 | - | 10 | 996 | 40146761 | ||
BC136760.1 | - | 10 | 996 | 9020750 | ||
BC144401.1 | - | 10 | 1054 |