Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130045.1 | NP_001123517.1 | 13 | 1468 | ||
NM_153254.2 | NP_694986.2 | 9 | 1275 | |||
XM_005244738.1 | XP_005244795.1 | 9 | 1302 | |||
XM_011541177.2 | XP_011539479.1 | 13 | 1545 | |||
XM_017000906.1 | XP_016856395.1 | 9 | 1546 | |||
XM_017000907.1 | XP_016856396.1 | 10 | 1627 | |||
XM_017000908.1 | XP_016856397.1 | 10 | 1627 | |||
XM_017000909.1 | XP_016856398.1 | 10 | 1627 | |||
XM_017000910.1 | XP_016856399.1 | 10 | 1627 | |||
XM_017000911.1 | XP_016856400.1 | 10 | 1627 | |||
XM_017000912.1 | XP_016856401.1 | 11 | 1399 | |||
GenBank mRNA | AK124125.1 | - | 13 | 1468 | ||
BC027945.1 | - | 13 | 1489 | |||
BC126152.1 | - | 9 | 1242 | 8991868 | ||
BC126154.1 | - | 9 | 1242 | 8991870 |