Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHYHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100876.1 | NP_001094346.1 | 12 | 1664 | ||
NM_001100877.1 | NP_001094347.1 | 9 | 709 | |||
NM_174933.3 | NP_777593.2 | 11 | 1642 | |||
GenBank mRNA | AK054994.1 | - | 13 | 1342 | ||
AK095000.1 | - | 12 | 1664 | |||
BC009373.2 | - | 11 | 1129 | 4121964 | ||
BC051300.1 | - | 9 | 681 | 4843267 | ||
BC053853.1 | - | 10 | 733 | 5209351 |