Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM86B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173804.4 | NP_776165.3 | 3 | 996 | ||
XM_017026554.1 | XP_016882043.1 | 2 | 769 | |||
GenBank mRNA | AK126545.1 | - | 1 | 1396 | ||
BC023000.1 | - | 3 | 996 | 5000011 | ||
BC033193.1 | - | 2 | 758 |