Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000806.5 | NP_000797.2 | 6 | 869 | ||
NM_001127643.1 | NP_001121115.1 | 6 | 756 | |||
NM_001127644.1 | NP_001121116.1 | 5 | 905 | |||
NM_001127645.1 | NP_001121117.1 | 5 | 493 | |||
NM_001127648.1 | NP_001121120.1 | 4 | 515 | |||
GenBank mRNA | AK223447.1 | - | 5 | 642 | ||
AK289792.1 | - | 6 | 869 | |||
AK295160.1 | - | 6 | 688 | |||
BC030696.1 | - | 5 | 470 | 4820972 | ||
DA228536.1 | - | 3 | 270 | |||
DA232066.1 | - | 1 | 165 | |||
DC307259.1 | - | 5 | 493 | |||
DC334834.1 | - | 4 | 515 | |||
X14766.1 | - | 5 | 615 |