Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174071.2 | NP_001167542.1 | 10, 11 | 1247 | ||
NM_001174072.2 | NP_001167543.1 | 10, 11 | 1247 | |||
NM_178276.6 | NP_840060.1 | 10, 11 | 1247 | |||
NR_126060.1 | 10, 11 | 1241 | ||||
NR_126061.1 | 10, 11 | 1247 | ||||
XM_011543304.1 | XP_011541606.1 | 10, 11 | 1212 | |||
XM_017009323.1 | XP_016864812.1 | 10, 11 | 1193 | |||
XM_017009324.1 | XP_016864813.1 | 10, 11 | 1212 | |||
XM_017009325.1 | XP_016864814.1 | 8, 9 | 848 | |||
XM_017009326.1 | XP_016864815.1 | 8, 9 | 848 | |||
GenBank mRNA | AF498273.1 | - | 10, 11 | 1217 | ||
AK297467.1 | - | 10, 11 | 1244 | |||
BC101280.2 | - | 10, 11 | 1179 | 40024444 | ||
BC101281.3 | - | 10, 11 | 1184 | 40024445 | ||
BC101283.2 | - | 10, 11 | 1184 | 40024448 |