Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025598.1 | NP_001020769.1 | 12 | 3063 | ||
NM_001287600.1 | NP_001274529.1 | 11 | 2918 | |||
NM_001287602.1 | NP_001274531.1 | 8 | 2532 | |||
NM_181720.2 | NP_859071.2 | 13 | 2430 | |||
XM_005245070.2 | XP_005245127.1 | 12 | 2892 | |||
XM_005245073.3 | XP_005245130.1 | 10 | 2816 | |||
XM_011509391.2 | XP_011507693.1 | 11 | 2894 | |||
XM_017000960.1 | XP_016856449.1 | 11 | 2661 | |||
GenBank mRNA | AK126163.1 | - | 8 | 2532 | ||
AK160366.1 | - | 12 | 3038 | |||
AL832852.1 | - | 10 | 2498 | |||
AL833477.1 | - | 1 | 1206 | |||
BC053688.1 | - | 13 | 2126 | 5213184 | ||
BX537846.1 | - | 10 | 3499 |