Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318146.1 | NP_001305075.1 | 1 | 639 | ||
NM_153271.1 | NP_695003.1 | 1 | 639 | |||
GenBank mRNA | AL833039.1 | - | 1 | 603 | ||
BC018775.1 | - | 1 | 639 | 4869639 | ||
EF219141.1 | - | 1 | 564 | |||
EF653821.1 | - | 1 | 805 |