Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012157.3 | NP_036289.3 | 7 | 461 | ||
XM_005265014.2 | XP_005265071.1 | 7 | 481 | |||
XM_005265015.3 | XP_005265072.1 | 7 | 701 | |||
XM_005265016.2 | XP_005265073.1 | 7 | 445 | |||
XM_005265017.3 | XP_005265074.1 | 7 | 477 | |||
XM_011533559.2 | XP_011531861.1 | 8 | 646 | |||
XM_017006092.1 | XP_016861581.1 | 8 | 538 | |||
XM_017006093.1 | XP_016861582.1 | 7 | 528 | |||
XM_017006094.1 | XP_016861583.1 | 8 | 561 | |||
GenBank mRNA | AF174589.1 | - | 8 | 432 | ||
AF176518.1 | - | 7 | 396 | |||
AF186273.1 | - | 7 | 411 | |||
AK001271.1 | - | 7 | 461 | |||
AK001438.1 | - | 7 | 429 | |||
AK294209.1 | - | 4 | 212 | |||
AK297534.1 | - | 7 | 445 | |||
AK299665.1 | - | 8 | 544 | |||
AK302218.1 | - | 9 | 689 | |||
AK303764.1 | - | 5 | 321 | |||
AK316383.1 | - | 8 | 601 | |||
AU124816.1 | - | 7 | 461 | |||
BC031556.1 | - | 7 | 405 | 5197151 | ||
CR457121.1 | - | 7 | 390 |