Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 11 | 732 | ||
NM_012157.3 | NP_036289.3 | 12 | 939 | |||
XM_005265014.2 | XP_005265071.1 | 12 | 959 | |||
XM_005265015.3 | XP_005265072.1 | 12 | 1179 | |||
XM_005265016.2 | XP_005265073.1 | 12 | 923 | |||
XM_005265017.3 | XP_005265074.1 | 12 | 955 | |||
XM_011533559.2 | XP_011531861.1 | 13 | 1124 | |||
XM_017006092.1 | XP_016861581.1 | 13 | 1016 | |||
XM_017006093.1 | XP_016861582.1 | 12 | 1006 | |||
XM_017006094.1 | XP_016861583.1 | 13 | 1039 | |||
GenBank mRNA | AF174589.1 | - | 13 | 910 | ||
AF176518.1 | - | 12 | 874 | |||
AF186273.1 | - | 12 | 889 | |||
AK001271.1 | - | 12 | 939 | |||
AK001438.1 | - | 12 | 907 | |||
AK294209.1 | - | 9 | 690 | |||
AK298967.1 | - | 11 | 701 | |||
AK299665.1 | - | 13 | 1022 | |||
AK302218.1 | - | 14 | 1167 | |||
AK303764.1 | - | 10 | 799 | |||
AK316229.1 | - | 12 | 960 | |||
AK316383.1 | - | 13 | 1079 | |||
AL049953.1 | - | 5 | 349 | |||
BC031556.1 | - | 12 | 883 | 5197151 | ||
CR457121.1 | - | 12 | 868 |