Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308053.1 | NP_001294982.1 | 5 | 546 | ||
NM_014353.4 | NP_055168.2 | 4 | 507 | |||
XM_011522448.1 | XP_011520750.1 | 4 | 533 | |||
XM_011522450.2 | XP_011520752.1 | 3 | 1324 | |||
GenBank mRNA | AB027137.1 | - | 3 | 178 | ||
AF498952.1 | - | 3 | 175 | |||
AK308560.1 | - | 6 | 603 | |||
AK309666.1 | - | 5 | 511 | |||
AK314110.1 | - | 5 | 546 | |||
AY646153.1 | - | 4 | 524 | |||
BC066913.1 | - | 4 | 513 | 5262795 |