Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024957.1 | NP_001020128.1 | 7, 8 | 770 | ||
NM_015399.3 | NP_056214.1 | 7, 8 | 770 | |||
GenBank mRNA | AF147350.1 | - | 1, 2 | 84 | ||
AF159141.1 | - | 7, 8 | 744 | |||
AK310991.1 | - | 7, 9 | 633 | |||
AK313773.1 | - | 7, 8 | 853 | |||
AL050008.1 | - | 5, 6 | 479 | |||
BC009834.2 | - | 7, 8 | 726 | 3959486 | ||
CB529276.1 | - | 3, 4 | 593 | |||
CR457173.1 | - | 6, 7 | 623 |