Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001556.2 | NP_001001556.1 | 2 | 330 | ||
NM_001289030.1 | NP_001275959.1 | 3 | 562 | |||
NM_001289031.1 | NP_001275960.1 | 2 | 415 | |||
NM_002044.3 | NP_002035.1 | 2 | 432 | |||
XM_005254279.3 | XP_005254336.1 | 2 | 221 | |||
XM_005254280.3 | XP_005254337.1 | 2 | 312 | |||
XM_005254284.3 | XP_005254341.1 | 2 | 312 | |||
XM_006720461.3 | XP_006720524.1 | 2 | 221 | |||
XM_006720462.3 | XP_006720525.1 | 2 | 202 | |||
XM_006720463.3 | XP_006720526.1 | 2 | 201 | |||
XM_006720464.3 | XP_006720527.1 | 2 | 201 | |||
XM_011521441.2 | XP_011519743.1 | 2 | 280 | |||
XM_017022062.1 | XP_016877551.1 | 2 | 202 | |||
XM_017022063.1 | XP_016877552.1 | 2 | 312 | |||
XM_017022065.1 | XP_016877554.1 | 2 | 332 | |||
XM_017022066.1 | XP_016877555.1 | 2 | 353 | |||
GenBank mRNA | AF461816.1 | - | 2 | 310 | ||
AK098049.1 | - | 2 | 171 | |||
AK301361.1 | - | 2 | 185 | |||
AK316440.1 | - | 3 | 296 | |||
AL832745.1 | - | 2 | 309 | |||
BC005141.2 | - | 2 | 200 | 3504930 | ||
BI766538.1 | - | 2 | 222 | |||
BQ921903.1 | - | 2 | 143 | |||
BT006901.1 | - | 2 | 125 | |||
DB457948.1 | - | 2 | 432 | |||
DC298656.1 | - | 2 | 422 | |||
DC390737.1 | - | 3 | 531 | |||
M84443.1 | - | 2 | 145 |