Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079880.1 | NP_001073349.1 | 8 | 1461 | ||
NM_001079881.1 | NP_001073350.1 | 8 | 1325 | |||
NM_001079882.1 | NP_001073351.1 | 6 | 1062 | |||
NM_016457.4 | NP_057541.2 | 7 | 1740 | |||
XM_005258716.2 | XP_005258773.2 | 6 | 718 | |||
GenBank mRNA | AF309082.1 | - | 7 | 1022 | ||
AK074673.1 | - | 6 | 701 | |||
AK075266.1 | - | 6 | 811 | |||
AK095884.1 | - | 8 | 1004 | |||
AK300339.1 | - | 8 | 1275 |