Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042469.1 | NP_001035934.2 | 3 | 360 | ||
NM_001042470.1 | NP_001035935.2 | 3 | 360 | |||
NM_001130069.2 | NP_001123541.1 | 3 | 360 | |||
NM_001146333.1 | NP_001139805.1 | 2 | 203 | |||
NM_015411.2 | NP_056226.2 | 3 | 360 | |||
XM_011515254.2 | XP_011513556.1 | 3 | 360 | |||
XM_017011939.1 | XP_016867428.1 | 3 | 360 | |||
XM_017011940.1 | XP_016867429.1 | 3 | 577 | |||
GenBank mRNA | AK075477.1 | - | 3 | 319 | ||
AK297042.1 | - | 3 | 276 | |||
AK297793.1 | - | 3 | 274 | |||
AK298605.1 | - | 3 | 300 | |||
AK300488.1 | - | 2 | 162 | |||
AK301627.1 | - | 3 | 277 | |||
AL050037.1 | - | 3 | 276 | |||
AY323911.1 | - | 3 | 272 | |||
AY359103.1 | - | 3 | 290 | |||
BC000224.1 | - | 2 | 95 | |||
BC006159.1 | - | 3 | 258 | |||
BC015600.2 | - | 3 | 257 | |||
BC084539.1 | - | 3 | 290 | 6599080 | ||
BC111092.1 | - | 3 | 313 | 40068651 | ||
BM423367.1 | - | 3 | 262 | |||
CF131206.1 | - | 3 | 321 | |||
CR936749.1 | - | 3 | 295 | |||
CR936757.1 | - | 3 | 303 | |||
CX164099.1 | - | 3 | 238 | |||
DA130450.1 | - | 3 | 338 | |||
DA917807.1 | - | 3 | 329 | |||
DA924174.1 | - | 3 | 361 | |||
DB010869.1 | - | 3 | 278 | |||
KU178582.1 | - | 2 | 185 | |||
KU178583.1 | - | 2 | 185 | |||
KU178584.1 | - | 2 | 185 | |||
KU178585.1 | - | 2 | 185 |