Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129727.2 | NP_001123199.1 | 13 | 1951 | ||
NM_001129728.1 | NP_001123200.1 | 12 | 1911 | |||
NM_001129729.2 | NP_001123201.1 | 12 | 1884 | |||
NM_001129731.2 | NP_001123203.1 | 10 | 1890 | |||
XM_011522985.2 | XP_011521287.1 | 12 | 1881 | |||
XM_011522986.2 | XP_011521288.1 | 12 | 2013 | |||
XM_011522987.2 | XP_011521289.1 | 13 | 1957 | |||
XM_011522988.2 | XP_011521290.1 | 12 | 1830 | |||
GenBank mRNA | AB197663.1 | - | 11 | 4147 | ||
AB197664.1 | - | 10 | 4314 | |||
AK024475.1 | - | 12 | 1846 | |||
AK304723.1 | - | 10 | 1302 | |||
AK308298.1 | - | 12 | 1820 | |||
AL117435.1 | - | 7 | 783 | |||
BC054486.1 | - | 10 | 1825 | 4820741 | ||
BC063501.1 | - | 12 | 1822 | 5527705 | ||
BC082974.1 | - | 13 | 1919 | 6291175 | ||
BC115390.1 | - | 9 | 1849 |